missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitropyridine 99.0+%, TCI America™

Catalog No. C10085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10085G 5 g
C100825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10085G Supplier TCI America Supplier No. C10085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5470-18-8 | C5H3ClN2O2 | 158.541 g/mol

Chemical Identifiers

CAS 5470-18-8
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.541
MDL Number MFCD00006232
InChI Key UUOLETYDNTVQDY-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
PubChem CID 79613
IUPAC Name 2-chloro-3-nitropyridine
SMILES C1=CC(=C(N=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 103°C
Color Yellow
Quantity 5 g
Formula Weight 158.54
Percent Purity ≥99.0% (GC)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-nitropyridine