missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitro-5-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. p-7125821
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30891G 1 g
C30895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30891G Supplier TCI America Supplier No. C30891G
Only null left

Chemical Identifiers

CAS 72587-15-6
Molecular Formula C6H2ClF3N2O2
Molecular Weight (g/mol) 226.539
MDL Number MFCD06255171
InChI Key DRPIKFKCAJGTJF-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-5-trifluoromethyl pyridine, 2-chloro-5-trifluoromethyl-3-nitropyridine, pyridine, 2-chloro-3-nitro-5-trifluoromethyl, 5-trifluoromethyl-3-nitro-2-chloropyridine, pubchem17529, acmc-209onn, abbypharma ap-11-5568, 2-chloro-3-nitro-5-trifluoromethyl-pyridine, 2-chloro3-nitro-5-trifluoromethyl pyridine
PubChem CID 4736950
IUPAC Name 2-chloro-3-nitro-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1[N+](=O)[O-])Cl)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 226.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-nitro-5-(trifluoromethyl)pyridine