missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitro-5-(trifluoromethyl)pyridine, 95%

Catalog No. p-7055741
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6439403 1 g
AAH6439406 5 g
AAH6439414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6439403 Supplier Thermo Scientific Chemicals Supplier No. H6439403
Only null left

CAS: 72587-15-6 | C6H2ClF3N2O2 | 226.539 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72587-15-6
Molecular Formula C6H2ClF3N2O2
Molecular Weight (g/mol) 226.539
MDL Number MFCD06255171
InChI Key DRPIKFKCAJGTJF-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-5-trifluoromethyl pyridine, 2-chloro-5-trifluoromethyl-3-nitropyridine, pyridine, 2-chloro-3-nitro-5-trifluoromethyl, 5-trifluoromethyl-3-nitro-2-chloropyridine, pubchem17529, acmc-209onn, abbypharma ap-11-5568, 2-chloro-3-nitro-5-trifluoromethyl-pyridine, 2-chloro3-nitro-5-trifluoromethyl pyridine
PubChem CID 4736950
IUPAC Name 2-chloro-3-nitro-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1[N+](=O)[O-])Cl)C(F)(F)F

Specifications

Color Colorless to Yellow
Quantity 1 g
Beilstein 7586778
Formula Weight 226.54
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-nitro-5-(trifluoromethyl)pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only