missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitro-4-picoline, 98%, Thermo Scientific™

Chemical Identifiers

CAS 23056-39-5
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD00012347
InChI Key JHARVUVBTAAPLA-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-4-picoline, pyridine, 2-chloro-4-methyl-3-nitro, 2-chloro-3-nitro-4-methylpyridine, 2-chloro-4-methyl-3-nitro-pyridine, pubchem6148, acmc-1cui4, 2-chloro-3-nitro4-picoline, ksc496c3n, 2-chlor-4-methyl-3-nitropyridin
PubChem CID 345363
IUPAC Name 2-chloro-4-methyl-3-nitropyridine
SMILES CC1=C(C(=NC=C1)Cl)[N+](=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC379450050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

23056-39-5
172.57
JHARVUVBTAAPLA-UHFFFAOYSA-N
345363
CC1=C(C(=NC=C1)Cl)[N+](=O)[O-]
C6H5ClN2O2
MFCD00012347
2-chloro-3-nitro-4-picoline, pyridine, 2-chloro-4-methyl-3-nitro, 2-chloro-3-nitro-4-methylpyridine, 2-chloro-4-methyl-3-nitro-pyridine, pubchem6148, acmc-1cui4, 2-chloro-3-nitro4-picoline, ksc496c3n, 2-chlor-4-methyl-3-nitropyridin
2-chloro-4-methyl-3-nitropyridine

Specifications

23056-39-5
100.0
Authentic
C6H5ClN2O2
2-chloro-3-nitro-4-picoline, pyridine, 2-chloro-4-methyl-3-nitro, 2-chloro-3-nitro-4-methylpyridine, 2-chloro-4-methyl-3-nitro-pyridine, pubchem6148, acmc-1cui4, 2-chloro-3-nitro4-picoline, ksc496c3n, 2-chlor-4-methyl-3-nitropyridin
CC1=C(C(=NC=C1)Cl)[N+](=O)[O-]
172.57
172.57
2-Chloro-3-nitro-4-picoline
97.5
113°C
97.5% min. (GC)
MFCD00012347
JHARVUVBTAAPLA-UHFFFAOYSA-N
2-chloro-4-methyl-3-nitropyridine
345363
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only