missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methylthiophene 98.0+%, TCI America™

Catalog No. p-7125180
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18015G 5 g
C180125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18015G Supplier TCI America Supplier No. C18015G

Chemical Identifiers

CAS 14345-97-2
Molecular Formula C5H5ClS
Molecular Weight (g/mol) 132.61
MDL Number MFCD00130083
InChI Key KQFADYXPELMVHE-UHFFFAOYSA-N
Synonym 2-chlor-3-methylthiophen, thiophene, 2-chloro-3-methyl, pubchem5164, acmc-209cqk, 2-chloro 3-methyl thiophene, 2-chloro-3-methyl thiophene, ksc174m0h, 2-chloro-3-methylthiophene
PubChem CID 84365
IUPAC Name 2-chloro-3-methylthiophene
SMILES CC1=C(Cl)SC=C1

Specifications

Color Yellow
Boiling Point 54°C
Quantity 5 g
UN Number 1993
Formula Weight 132.61
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-methylthiophene