missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methylpyridine 98.0+%, TCI America™

Catalog No. C18855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18855G 5 g
C188525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18855G Supplier TCI America Supplier No. C18855G

Chemical Identifiers

CAS 18368-76-8
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
InChI Key RKVUCIFREKHYTL-UHFFFAOYSA-N
PubChem CID 87603
IUPAC Name 2-chloro-3-methylpyridine
SMILES CC1=C(N=CC=C1)Cl

Specifications

Color Yellow
Boiling Point 193°C
Quantity 5 g
Synonym 2-chloro-3-picoline, 2-chloro 3-methylpyridine, 3-picoline, 2-chloro, pyridine, 2-chloro-3-methyl, 2-chloro-3-methyl-pyridine, pyridine,2-chloro-3-methyl, pubchem2074, 2-chloro-methyl-pyridine, 2-chloro3-methyl-pyridine
Formula Weight 127.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-methylpyridine