missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methyl-5-nitropyridine 98.0+%, TCI America™

Catalog No. p-7125238
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20201G 1 g
C20205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20201G Supplier TCI America Supplier No. C20201G
Only null left

Chemical Identifiers

CAS 22280-56-4
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
InChI Key OSIOIGXJUZTWRI-UHFFFAOYSA-N
Synonym 2-chloro-5-nitro-3-picoline, pyridine, 2-chloro-3-methyl-5-nitro, 2-chloro-3-methyl-5-nitro pyridine, 2-chloro-3-methyl-5-nitro-pyridine, 2-chloro-5-nitro-3-methylpyridine, pubchem10621, acmc-209fub, ksc207a2n, osioigxjuztwri-uhfffaoysa
PubChem CID 89648
IUPAC Name 2-chloro-3-methyl-5-nitropyridine
SMILES CC1=CC(=CN=C1Cl)[N+](=O)[O-]

Specifications

Melting Point 49°C
Boiling Point 145°C
Quantity 1 g
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-methyl-5-nitropyridine