missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-3-methyl-5-nitropyridine, 97%, Thermo Scientific™

Catalog No. BTB09487DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB09487DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB09487DA Supplier Thermo Scientific Chemicals Supplier No. BTB09487DA
Only null left

Chemical Identifiers

CAS 22280-56-4
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00173686
InChI Key OSIOIGXJUZTWRI-UHFFFAOYSA-N
Synonym 2-chloro-5-nitro-3-picoline, pyridine, 2-chloro-3-methyl-5-nitro, 2-chloro-3-methyl-5-nitro pyridine, 2-chloro-3-methyl-5-nitro-pyridine, 2-chloro-5-nitro-3-methylpyridine, pubchem10621, acmc-209fub, ksc207a2n, osioigxjuztwri-uhfffaoysa
PubChem CID 89648
IUPAC Name 2-chloro-3-methyl-5-nitropyridine
SMILES CC1=CC(=CN=C1Cl)[N+](=O)[O-]

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 172.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-methyl-5-nitropyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.