missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-iodopyridine 98.0+%, TCI America™

Catalog No. C21935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21935G 5 g
C21931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21935G Supplier TCI America Supplier No. C21935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78607-36-0 | C5H3ClIN | 239.44 g/mol

Chemical Identifiers

CAS 78607-36-0
Molecular Formula C5H3ClIN
Molecular Weight (g/mol) 239.44
MDL Number MFCD00661298
InChI Key OHWSWGXNZDSHLM-UHFFFAOYSA-N
Synonym 2-chloro-3-iodo-pyridine, pyridine, 2-chloro-3-iodo, 3-iodo-2-chloropyridine, zlchem 306, pubchem1188, acmc-209pes, 2-chloro-3-iodopyridine, ksc378k0b, 2-chloranyl-3-iodanyl-pyridine, 2-choro-3-iodopyridine
PubChem CID 2735743
IUPAC Name 2-chloro-3-iodopyridine
SMILES C1=CC(=C(N=C1)Cl)I

Specifications

Melting Point 95°C
Quantity 5 g
Formula Weight 239.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-iodopyridine