missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-iodopyridine, 98+%, Thermo Scientific™

Catalog No. AC379510050
Change view
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC379510050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC379510050 Supplier Thermo Scientific Chemicals Supplier No. 379510050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78607-36-0
Molecular Formula C5H3ClIN
Molecular Weight (g/mol) 239.44
MDL Number MFCD00661298
InChI Key OHWSWGXNZDSHLM-UHFFFAOYSA-N
Synonym 2-chloro-3-iodo-pyridine, pyridine, 2-chloro-3-iodo, 3-iodo-2-chloropyridine, zlchem 306, pubchem1188, acmc-209pes, 2-chloro-3-iodopyridine, ksc378k0b, 2-chloranyl-3-iodanyl-pyridine, 2-choro-3-iodopyridine
PubChem CID 2735743
IUPAC Name 2-chloro-3-iodopyridine
SMILES C1=CC(=C(N=C1)Cl)I

Specifications

CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5g
Solubility Information Solubility in water: insoluble
Formula Weight 239.44
Percent Purity 98+%
Chemical Name or Material 2-Chloro-3-iodopyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only