missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-hydroxypyridine 98.0+%, TCI America™

$59.69 - $119.36

Chemical Identifiers

CAS 6636-78-8
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD00006235
InChI Key RSOPTYAZDFSMTN-UHFFFAOYSA-N
Synonym 2-chloro-3-hydroxypyridine, 2-chloro-3-pyridinol, 3-pyridinol, 2-chloro, unii-vy212q0s3j, 2-chloro-3-hydroxy pyridine, 2-chloro-3-hydroxypyridine 2-chloro-3-pyridinol, 2-chlorpyridin-3-ol, pubchem2577, 2-chloro-pyridin-3-ol, 2-chloro-pyridine-3-ol
PubChem CID 81136
IUPAC Name 2-chloropyridin-3-ol
SMILES OC1=CC=CN=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C115210G
View Documents
TCI America
C115210G
10 g
Each for $59.69
Only null left
 
C115225G
View Documents
TCI America
C115225G
25 g
Each for $119.36
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6636-78-8
129.54
RSOPTYAZDFSMTN-UHFFFAOYSA-N
81136
OC1=CC=CN=C1Cl
C5H4ClNO
MFCD00006235
2-chloro-3-hydroxypyridine, 2-chloro-3-pyridinol, 3-pyridinol, 2-chloro, unii-vy212q0s3j, 2-chloro-3-hydroxy pyridine, 2-chloro-3-hydroxypyridine 2-chloro-3-pyridinol, 2-chlorpyridin-3-ol, pubchem2577, 2-chloro-pyridin-3-ol, 2-chloro-pyridine-3-ol
2-chloropyridin-3-ol

Specifications

6636-78-8
C5H4ClNO
10 g
RSOPTYAZDFSMTN-UHFFFAOYSA-N
2-chloropyridin-3-ol
81136
≥98.0% (GC,T)
2-Chloro-3-hydroxypyridine
167°C
MFCD00006235
2-chloro-3-hydroxypyridine, 2-chloro-3-pyridinol, 3-pyridinol, 2-chloro, unii-vy212q0s3j, 2-chloro-3-hydroxy pyridine, 2-chloro-3-hydroxypyridine 2-chloro-3-pyridinol, 2-chlorpyridin-3-ol, pubchem2577, 2-chloro-pyridin-3-ol, 2-chloro-pyridine-3-ol
OC1=CC=CN=C1Cl
129.54
129.54
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UU7707000

TSCA : No

SDS
Documents

Documents