missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-hydroxypyridine 98.0+%, TCI America™

Catalog No. C115225G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C115225G 25 g
C115210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C115225G Supplier TCI America Supplier No. C115225G

Chemical Identifiers

CAS 6636-78-8
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD00006235
InChI Key RSOPTYAZDFSMTN-UHFFFAOYSA-N
Synonym 2-chloro-3-hydroxypyridine, 2-chloro-3-pyridinol, 3-pyridinol, 2-chloro, unii-vy212q0s3j, 2-chloro-3-hydroxy pyridine, 2-chloro-3-hydroxypyridine 2-chloro-3-pyridinol, 2-chlorpyridin-3-ol, pubchem2577, 2-chloro-pyridin-3-ol, 2-chloro-pyridine-3-ol
PubChem CID 81136
IUPAC Name 2-chloropyridin-3-ol
SMILES OC1=CC=CN=C1Cl

Specifications

Melting Point 167°C
Quantity 25 g
Formula Weight 129.54
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-hydroxypyridine
RTECSNumber UU7707000
TSCA No