missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3-hydroxypyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 167°C |
| Quantity | 10 g |
| Formula Weight | 129.54 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Chloro-3-hydroxypyridine |
Safety and Handling
| RTECSNumber | UU7707000 |
| TSCA | No |