missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoropyridine-6-carboxylic acid, 98%

CAS: 860296-24-8 | C6H3ClFNO2 | 175.543 g/mol

$106.27 - $356.59

Chemical Identifiers

CAS 860296-24-8
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.543
MDL Number MFCD13185819
InChI Key JHFKXHUKNYFWJM-UHFFFAOYSA-N
Synonym 6-chloro-5-fluoropicolinic acid, 2-chloro-3-fluoropyridine-6-carboxylicacid, 2-chloro-3-fluoropyridine-6-carboxylic acid, 6-chloro-5-fluoro-pyridine-2-carboxylic acid, 2-chloro-3-fluoro-6-carboxypyridine, 2-chloro-3-fluoro-6-pyridinecarboxylic acid
PubChem CID 11412672
IUPAC Name 6-chloro-5-fluoropyridine-2-carboxylic acid
SMILES C1=CC(=NC(=C1F)Cl)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6473403
View Documents
Thermo Scientific Chemicals
H6473403
1 g
Each for $106.27
Only null left
 
AAH6473406
View Documents
Thermo Scientific Chemicals
H6473406
5 g
Each for $356.59
Only null left
 
Description

Description

It is an important intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

860296-24-8
175.543
JHFKXHUKNYFWJM-UHFFFAOYSA-N
11412672
C1=CC(=NC(=C1F)Cl)C(=O)O
C6H3ClFNO2
MFCD13185819
6-chloro-5-fluoropicolinic acid, 2-chloro-3-fluoropyridine-6-carboxylicacid, 2-chloro-3-fluoropyridine-6-carboxylic acid, 6-chloro-5-fluoro-pyridine-2-carboxylic acid, 2-chloro-3-fluoro-6-carboxypyridine, 2-chloro-3-fluoro-6-pyridinecarboxylic acid
6-chloro-5-fluoropyridine-2-carboxylic acid

Specifications

860296-24-8
C6H3ClFNO2
1 g
6-chloro-5-fluoropicolinic acid, 2-chloro-3-fluoropyridine-6-carboxylicacid, 2-chloro-3-fluoropyridine-6-carboxylic acid, 6-chloro-5-fluoro-pyridine-2-carboxylic acid, 2-chloro-3-fluoro-6-carboxypyridine, 2-chloro-3-fluoro-6-pyridinecarboxylic acid
JHFKXHUKNYFWJM-UHFFFAOYSA-N
6-chloro-5-fluoropyridine-2-carboxylic acid
11412672
98%
2-Chloro-3-fluoropyridine-6-carboxylic acid
White
MFCD13185819
10063182
Slightly soluble in water.
C1=CC(=NC(=C1F)Cl)C(=O)O
175.543
175.54
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only