missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoro-5-methylpyridine 98.0+%, TCI America™

Catalog No. p-7125914
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
C32751G 1 g
C32755G 5 g
2 options

Catalog No. C32751G

Supplier: TCI America C32751G

Only null left

Chemical Identifiers

CAS 34552-15-3
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD06658238
InChI Key XZJURWKNRAGMCG-UHFFFAOYSA-N
Synonym 2-chloro-3-fluoro-5-picoline, 2-chloro-3-fluoro-5-methyl-pyridine, 6-chloro-5-fluoro-3-picoline, pyridine, 2-chloro-3-fluoro-5-methyl, 2-chloro-3-fluoro-5-methyl pyridine, pubchem14231, ksc495q2t, 6-chloro-5-fluoro-3-methylpyridine
PubChem CID 13541365
IUPAC Name 2-chloro-3-fluoro-5-methylpyridine
SMILES CC1=CC(F)=C(Cl)N=C1

Specifications

Color White-Yellow
Boiling Point 92°C
Quantity 1 g
Formula Weight 145.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-3-fluoro-5-methylpyridine
TSCA No
Recommended Storage Refrigerator