missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoro-5-methylpyridine, 95%, Thermo Scientific Chemicals

Catalog No. p-7054081
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
AAH6197206 5 g
1 options

Catalog No. AAH6197206

Supplier: Thermo Scientific Chemicals H6197206

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34552-15-3
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD06658238
InChI Key XZJURWKNRAGMCG-UHFFFAOYSA-N
Synonym 2-chloro-3-fluoro-5-picoline, 2-chloro-3-fluoro-5-methyl-pyridine, 6-chloro-5-fluoro-3-picoline, pyridine, 2-chloro-3-fluoro-5-methyl, 2-chloro-3-fluoro-5-methyl pyridine, pubchem14231, ksc495q2t, 6-chloro-5-fluoro-3-methylpyridine
PubChem CID 13541365
IUPAC Name 2-chloro-3-fluoro-5-methylpyridine
SMILES CC1=CC(F)=C(Cl)N=C1

Specifications

Color White
Quantity 5 g
Formula Weight 145.56
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Chloro-3-fluoro-5-methylpyridine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only