missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-ethylbenzoxazolium Tetrafluoroborate 97.0+%, TCI America™

Catalog No. C09055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09055G 5 g
C090525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09055G Supplier TCI America Supplier No. C09055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63212-53-3
Molecular Formula C9H9BClF4NO
Molecular Weight (g/mol) 269.431
MDL Number MFCD00011943
InChI Key MLQZANYFUYLJRK-UHFFFAOYSA-N
Synonym 2-chloro-3-ethylbenzoxazolium tetrafluoroborate, 2-chloro-3-ethylbenzo d oxazol-3-ium tetrafluoroborate, 2-chloro-3-ethyl-1,3-benzoxazol-3-ium tetrafluoroborate, 2-chloro-3-ethyl-benzooxazole; tetrafluoroboron, acmc-209ndy, 3-ethyl-2-chlorobenzoxazolium tetrafluoroborate, 2-chloro-3-ethylbenzoxazo-lium tetrafluoroborate
PubChem CID 2724240
IUPAC Name 2-chloro-3-ethyl-1,3-benzoxazol-3-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CC[N+]1=C(OC2=CC=CC=C21)Cl

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 269.43
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-ethylbenzoxazolium Tetrafluoroborate
TSCA No
Recommended Storage Freezer