missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3-cyanopyridine 97.0+%, TCI America™
$52.19 - $607.68
Chemical Identifiers
| CAS | 6602-54-6 |
|---|---|
| Molecular Formula | C6H3ClN2 |
| Molecular Weight (g/mol) | 138.554 |
| MDL Number | MFCD00014628 |
| InChI Key | JAUPUQRPBNDMDT-UHFFFAOYSA-N |
| Synonym | 2-chloro-3-cyanopyridine, 2-chloronicotinonitrile, 2-chloro-3-pyridinecarbonitrile, 3-cyano-2-chloropyridine, 3-pyridinecarbonitrile, 2-chloro, 2-chloro-nicotinonitrile, 2-chloronicotinitrile, 2-chloro-3-cyano-pyridine, 2-chloro-3-cyanopyridine 2-chloronicotinonitrile, 2chloronicotinonitrile |
| PubChem CID | 81079 |
| IUPAC Name | 2-chloropyridine-3-carbonitrile |
| SMILES | C1=CC(=C(N=C1)Cl)C#N |
Chemical Identifiers
| 6602-54-6 | |
| 138.554 | |
| JAUPUQRPBNDMDT-UHFFFAOYSA-N | |
| 81079 | |
| C1=CC(=C(N=C1)Cl)C#N |
| C6H3ClN2 | |
| MFCD00014628 | |
| 2-chloro-3-cyanopyridine, 2-chloronicotinonitrile, 2-chloro-3-pyridinecarbonitrile, 3-cyano-2-chloropyridine, 3-pyridinecarbonitrile, 2-chloro, 2-chloro-nicotinonitrile, 2-chloronicotinitrile, 2-chloro-3-cyano-pyridine, 2-chloro-3-cyanopyridine 2-chloronicotinonitrile, 2chloronicotinonitrile | |
| 2-chloropyridine-3-carbonitrile |
Specifications
| 6602-54-6 | |
| White | |
| MFCD00014628 | |
| 3439 | |
| JAUPUQRPBNDMDT-UHFFFAOYSA-N | |
| 2-chloropyridine-3-carbonitrile | |
| 81079 | |
| ≥97.0% (GC) | |
| 2-Chloro-3-cyanopyridine |
| 106°C | |
| C6H3ClN2 | |
| 25 g | |
| 2-chloro-3-cyanopyridine, 2-chloronicotinonitrile, 2-chloro-3-pyridinecarbonitrile, 3-cyano-2-chloropyridine, 3-pyridinecarbonitrile, 2-chloro, 2-chloro-nicotinonitrile, 2-chloronicotinitrile, 2-chloro-3-cyano-pyridine, 2-chloro-3-cyanopyridine 2-chloronicotinonitrile, 2chloronicotinonitrile | |
| C1=CC(=C(N=C1)Cl)C#N | |
| 138.554 | |
| 138.55 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6782
TSCA : No