missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3-cyanopyridine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 106°C |
| Color | White |
| Quantity | 500 g |
| UN Number | 3439 |
| Formula Weight | 138.55 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Chloro-3-cyanopyridine |
Safety and Handling
| EINECSNumber | (5)-6782 |
| TSCA | No |