missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(chloromethyl)quinoline 98.0+%, TCI America™

Catalog No. C28615G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28615G 5 g
C28611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28615G Supplier TCI America Supplier No. C28615G
Only null left

Chemical Identifiers

CAS 90097-52-2
Molecular Formula C10H7Cl2N
Molecular Weight (g/mol) 212.073
MDL Number MFCD05865134
InChI Key UOIJHUIOTFDGNA-UHFFFAOYSA-N
Synonym 2-chloro-3-chloromethyl quinoline, quinoline, 2-chloro-3-chloromethyl, 2-chloranyl-3-chloromethyl quinoline
PubChem CID 2063384
IUPAC Name 2-chloro-3-(chloromethyl)quinoline
SMILES C1=CC=C2C(=C1)C=C(C(=N2)Cl)CCl

Specifications

Melting Point 117°C
Color Yellow
Quantity 5 g
UN Number 3261
Formula Weight 212.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-(chloromethyl)quinoline