missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-dinitropyridine 98.0+%, TCI America™

Catalog No. p-7124698
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09431G 1 g
C09435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09431G Supplier TCI America Supplier No. C09431G
Only null left

Chemical Identifiers

CAS 2578-45-2
Molecular Formula C5H2ClN3O4
Molecular Weight (g/mol) 203.538
MDL Number MFCD00006233
InChI Key QLHVJBXAQWPEDI-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-3,5-dinitro, 2-chloro-3,5-dinitro-pyridine, pubchem6103, acmc-209gmh, 2-chloro3,5-dinitropyridine, 2-chloro-3,5-dinitropyridine
PubChem CID 75738
IUPAC Name 2-chloro-3,5-dinitropyridine
SMILES C1=C(C=NC(=C1[N+](=O)[O-])Cl)[N+](=O)[O-]

Specifications

Melting Point 66°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 203.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3,5-dinitropyridine
RTECSNumber US6504750
TSCA Yes