missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3,5-dinitropyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 66°C |
| Color | Yellow |
| Quantity | 1 g |
| UN Number | 2811 |
| Formula Weight | 203.54 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Chloro-3,5-dinitropyridine |
Safety and Handling
| RTECSNumber | US6504750 |
| TSCA | Yes |