missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-dinitropyridine, 98+%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 208190010

Catalog No. AC208190010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

 

Specifications

Specifications

2-Chloro-3,5-dinitropyridine
2578-45-2
100.0
98% min. (GC)
C5H2ClN3O4
MFCD00006233
20,III,2536
QLHVJBXAQWPEDI-UHFFFAOYSA-N
2-chloro-3,5-dinitropyridine
75738
98+%
99%
98.5
Authentic
Glass bottle
(O2H)(C5H2N)Cl
1g
pyridine, 2-chloro-3,5-dinitro, 2-chloro-3,5-dinitro-pyridine, pubchem6103, acmc-209gmh, 2-chloro3,5-dinitropyridine, 2-chloro-3,5-dinitropyridine
C1=C(C=NC(=C1[N+](=O)[O-])Cl)[N+](=O)[O-]
203.54
203.54

Chemical Identifiers

2578-45-2
203.54
QLHVJBXAQWPEDI-UHFFFAOYSA-N
75738
C1=C(C=NC(=C1[N+](=O)[O-])Cl)[N+](=O)[O-]
C5H2ClN3O4
MFCD00006233
pyridine, 2-chloro-3,5-dinitro, 2-chloro-3,5-dinitro-pyridine, pubchem6103, acmc-209gmh, 2-chloro3,5-dinitropyridine, 2-chloro-3,5-dinitropyridine
2-chloro-3,5-dinitropyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing

GHS Signal Word: Danger

EINECSNumber : 219-937-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only