missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,4-dimethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7125230
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19975G 5 g
C199725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19975G Supplier TCI America Supplier No. C19975G
Only null left

Chemical Identifiers

CAS 5417-17-4
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
InChI Key SAWHDJTZESXNMM-UHFFFAOYSA-N
PubChem CID 79445
IUPAC Name 2-chloro-3,4-dimethoxybenzaldehyde
SMILES COC1=C(C(=C(C=C1)C=O)Cl)OC

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 5 g
Synonym 2-chloroveratraldehyde, 2-chloro-3,4-dimethoxy-benzaldehyde, benzaldehyde, 2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxy benzaldehyde, chloroveratraldehyde, pubchem7759, acmc-1aumn, 2-chloro-3,4 dimethoxybenzaldehyde, benzaldehyde,2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxybenzaldehyde
Formula Weight 200.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3,4-dimethoxybenzaldehyde