missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-oxo-1,3,2-dioxaphospholane 95.0+%, TCI America™

Catalog No. C12505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12505G 5 g
C125025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12505G Supplier TCI America Supplier No. C12505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6609-64-9
Molecular Formula C2H4ClO3P
Molecular Weight (g/mol) 142.475
MDL Number MFCD00043138
InChI Key SBMUNILHNJLMBF-UHFFFAOYSA-N
Synonym 2-chloro-2-oxo-1,3,2-dioxaphospholane, ethylene chlorophosphate, 2-chloro-1,3,2-dioxaphospholane-2-oxide, 2-chloro-1,3,2-dioxaphospholane 2-oxide, ethylene glycol chlorophosphate, soqdebhb^bsvusup@, 2-chloro-1,3,2??-dioxaphospholan-2-one, 2-chlor-1,3,2-dioxaphospholan-2-oxid, acmc-1b2u0, ethylene cycl-chlorophosphate
PubChem CID 81087
IUPAC Name 2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide
SMILES C1COP(=O)(O1)Cl

Specifications

Color Yellow
Boiling Point 91°C
Quantity 5 g
UN Number 3265
Formula Weight 142.47
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-oxo-1,3,2-dioxaphospholane
EINECSNumber (9)-0448
TSCA No
Recommended Storage Freezer