missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylpropane 98.0+%, TCI America™

Catalog No. p-7124281
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
C020825ML 25 mL
C0208500ML 500 mL
2 options

Catalog No. C020825ML

Supplier: TCI America C020825ML

Only null left

Chemical Identifiers

CAS 507-20-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000816
InChI Key NBRKLOOSMBRFMH-UHFFFAOYSA-N
Synonym tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
PubChem CID 10486
IUPAC Name 2-chloro-2-methylpropane
SMILES CC(C)(C)Cl

Specifications

Color Colorless
Boiling Point 52°C
Quantity 25 mL
UN Number 1127
Formula Weight 92.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylpropane
EINECSNumber (2)-0060
RTECSNumber TX5040000
TSCA Yes