missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylpropane 98.0+%, TCI America™

Catalog No. C020825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C020825ML 25 mL
C0208500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C020825ML Supplier TCI America Supplier No. C020825ML

Chemical Identifiers

CAS 507-20-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000816
InChI Key NBRKLOOSMBRFMH-UHFFFAOYSA-N
Synonym tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
PubChem CID 10486
IUPAC Name 2-chloro-2-methylpropane
SMILES CC(C)(C)Cl

Specifications

Color Colorless
Boiling Point 52°C
Quantity 25 mL
UN Number 1127
Formula Weight 92.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylpropane
EINECSNumber (2)-0060
RTECSNumber TX5040000
TSCA Yes