Learn More
2-Chloro-2-methylpropane, 99%
2-Chloro-2-methylpropane, 99%, C4H9Cl, CAS Number-507-20-0, 2-methyl-2-chloropropane, propane, 2-chloro-2-methyl, 2-chloroisobutane, tert-butylchloride, t-butylchloride, chlorotrimethylmethane, trimethylchloromethane, tert-butyl chloride, t-butyl chloride, 2-methyl-2-propyl chloride, 500mL | CAS: 507-20-0 | C4H9Cl | 92.57 g/mol
Supplier: Thermo Scientific Chemicals 109500025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Chloro-2-methylpropane | |
| 507-20-0 | |
| 100.0 | |
| 0.8500g/mL | |
| −27°C | |
| 98.5% min. (GC) | |
| C4H9Cl | |
| (CH3)3CCl | |
| 2.5 L | |
| 02,46 | |
| 15, 1563 | |
| Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether | |
| CC(C)(C)Cl | |
| 92.57 | |
| 0.51 mPa.s (20°C) | |
| 99% |
| 99% | |
| 98.5 | |
| -25.0°C | |
| 51.0°C to 52.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3838 to 1.3858 | |
| MFCD00000816 | |
| 01, 125 | |
| 0.85 | |
| tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride | |
| NBRKLOOSMBRFMH-UHFFFAOYSA-N | |
| 2-chloro-2-methylpropane | |
| 10486 | |
| 92.57 | |
| Liquid |
Chemical Identifiers
| 507-20-0 | |
| 92.57 | |
| NBRKLOOSMBRFMH-UHFFFAOYSA-N | |
| 10486 | |
| CC(C)(C)Cl |
| C4H9Cl | |
| MFCD00000816 | |
| tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride | |
| 2-chloro-2-methylpropane |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
GHS Signal Word: Danger
EINECSNumber : 208-066-4
RTECSNumber : TX5040000
TSCA : TSCA
RUO – Research Use Only