missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylnonane, 90+%

Catalog No. p-7058985
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4378406 5 g
AA4378414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4378406 Supplier Thermo Scientific Chemicals Supplier No. 04378406
Only null left

CAS: 4325-50-2 | C10H21Cl | 176.728 g/mol

It is used in the photoelectron spectroscopy of C60Fn- and C60Fm2 in the gas phase and the generation and characterization of C1-C60F47- and D2-C60F44 in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4325-50-2
Molecular Formula C10H21Cl
Molecular Weight (g/mol) 176.728
MDL Number MFCD00060781
InChI Key QDJXQCKHBZEIJM-UHFFFAOYSA-N
Synonym dimethyloctyl chloride, acmc-20ao0l, 2-chloro-2-methyl-nonane, nonane,2-chloro-2-methyl
PubChem CID 138189
IUPAC Name 2-chloro-2-methylnonane
SMILES CCCCCCCC(C)(C)Cl

Specifications

Color Colorless
Quantity 5 g
UN Number UN3082
Formula Weight 176.73
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylnonane
Hazard Category H351
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only