missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-ethoxyacetic acid ethyl ester, 90%, remainder mainly ethyl diethoxyacetat, Thermo Scientific Chemicals

Catalog No. AC279870500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC279870500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC279870500 Supplier Thermo Scientific Chemicals Supplier No. 279870500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34006-60-5 | Molecular Formula: C6H11ClO3 | Molecular Weight: 166.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34006-60-5
Molecular Formula C6H11ClO3
Molecular Weight (g/mol) 166.60
MDL Number MFCD00043953
InChI Key WDCHMODANTVRTF-UHFFFAOYNA-N
Synonym 2-chloro-2-ethoxyacetic acid ethyl ester, ethyl 2-chloro-2-ethoxy-acetate, ethyl2-chloro-2-ethoxyacetate, ethyl chloro ethoxy acetate, chloro ethoxy acetic acid ethyl ester, ethyl alpha-chloro-alpha-ethoxyacetate, ethyl alpha-chloro-alpha-ethoxy-acetate, 2-chloro-2-ethoxy-acetic acid ethyl ester, acetic acid,2-chloro-2-ethoxy-, ethyl ester
PubChem CID 2724873
IUPAC Name ethyl 2-(1-chloroethoxy)acetate
SMILES CCOC(=O)COC(C)Cl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Brown-Orange to Colorless
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4307
Linear Formula C2H5O(Cl)CHC(O)OC2H5
Quantity 50 g
Formula Weight 166.6
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-ethoxyacetic acid ethyl ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 251-786-9
TSCA TSCA

RUO – Research Use Only