missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2',4'-difluoroacetophenone 98.0+%, TCI America™

Catalog No. C20305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20305G 5 g
C203025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20305G Supplier TCI America Supplier No. C20305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51336-94-8
Molecular Formula C8H5ClF2O
Molecular Weight (g/mol) 190.574
InChI Key UENGBOCGGKLVJJ-UHFFFAOYSA-N
Synonym 2-chloro-2',4'-difluoroacetophenone, 2-chloro-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl chloride, 2-chloro-2,4-difluoroacetophenone, 2',4'-difluoro-alfa-chloroacetophenone, 2-chloro-1-2,4-difluorophenyl ethan-1-one, 2',4'-difluoro-2-chloroacetophenone, 1-2,4-difluorophenyl-2-chloroethan-1-one, 2',4'-difluorophenacyl chloride, 2'-chloro-2,4-difluoroacetophenone
PubChem CID 588083
IUPAC Name 2-chloro-1-(2,4-difluorophenyl)ethanone
SMILES C1=CC(=C(C=C1F)F)C(=O)CCl

Specifications

Melting Point 50°C
Quantity 5 g
UN Number 3261
Formula Weight 190.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-2′,4′-difluoroacetophenone