missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1H-imidazole 98.0+%, TCI America™

$591.03 - $591.03

Chemical Identifiers

CAS 16265-04-6
Molecular Formula C3H3ClN2
Molecular Weight (g/mol) 102.52
MDL Number MFCD02179530
InChI Key OCVXSFKKWXMYPF-UHFFFAOYSA-N
PubChem CID 2773328
ChEBI CHEBI:52721
IUPAC Name 2-chloro-1H-imidazole
SMILES ClC1=NC=CN1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C26851G
View Documents
TCI America
C26851G
1 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

16265-04-6
102.52
OCVXSFKKWXMYPF-UHFFFAOYSA-N
CHEBI:52721
ClC1=NC=CN1
C3H3ClN2
MFCD02179530
2773328
2-chloro-1H-imidazole

Specifications

16265-04-6
White-Yellow
MFCD02179530
2-chloroimidazole, chloroimidazole, sftheadililuh@, pubchem7597, 2-chlor-1h-imidazol, zlchem 1064, acmc-209dos, 1h-imidazole,2-chloro, 2-chloranyl-1h-imidazole, 1h-imidazole, 2-chloro
ClC1=NC=CN1
102.52
CHEBI:52721
≥98.0% (GC)
2-Chloro-1H-imidazole
168°C
C3H3ClN2
1 g
OCVXSFKKWXMYPF-UHFFFAOYSA-N
2-chloro-1H-imidazole
2773328
102.52
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'