missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine, 98%

Catalog No. AAJ64066MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64066MD 250 mg
AAJ6406603 1 g
AAJ6406606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ64066MD Supplier Thermo Scientific Chemicals Supplier No. J64066MD

A tricyclic norepinephrine uptake inhibitor | CAS: 14028-44-5 | C17H16ClN3O | 313.785 g/mol

A tricyclic norepinephrine uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14028-44-5
Molecular Formula C17H16ClN3O
Molecular Weight (g/mol) 313.785
MDL Number MFCD00069210
InChI Key QWGDMFLQWFTERH-UHFFFAOYSA-N
Synonym amoxapine, asendin, demolox, moxadil, amoxan, amoxapinum, amoxapina, amoxepine, asendis, desmethylloxapin
PubChem CID 2170
ChEBI CHEBI:2675
IUPAC Name 8-chloro-6-piperazin-1-ylbenzo[b][1,4]benzoxazepine
SMILES C1CN(CCN1)C2=NC3=CC=CC=C3OC4=C2C=C(C=C4)Cl

Specifications

Color White
Quantity 250 mg
Merck Index 14,576
Solubility Information Soluble in methanol,DMSO or chloroform.
Formula Weight 313.78
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 237-867-1
RTECSNumber HQ4025500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only