missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-propene 97.0+%, TCI America™

Catalog No. C111210ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C111210ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C111210ML Supplier TCI America Supplier No. C111210ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 557-98-2
Molecular Formula C3H5Cl
Molecular Weight (g/mol) 76.523
MDL Number MFCD00000859
InChI Key PNLQPWWBHXMFCA-UHFFFAOYSA-N
Synonym 2-chloropropene, 1-propene, 2-chloro, isopropenyl chloride, 2-chloropropylene, 2-chloro-1-propene, beta-chloropropene, beta-chloropropylene, propene, 2-chloro, unii-50txn55e4a, 2-chloro propylene
PubChem CID 11203
IUPAC Name 2-chloroprop-1-ene
SMILES CC(=C)Cl

Specifications

Color Yellow
Boiling Point 23°C
Quantity 10 mL
UN Number 2456
Formula Weight 76.52
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-1-propene
EINECSNumber (2)-0123
RTECSNumber UC7200000
TSCA Yes
Recommended Storage Refrigerator