missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Chloro-1-phenylethanol, 97%

Catalog No. AAL1453806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1453806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1453806 Supplier Thermo Scientific Chemicals Supplier No. L1453806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1674-30-2 | C8H9ClO | 156.609 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1674-30-2
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD01075658
InChI Key XWCQSILTDPAWDP-UHFFFAOYSA-N
Synonym styrene chlorohydrin, 1-phenyl-2-chloroethanol, +/--2-chloro-1-phenylethanol, 2-chloro-1-phenyl-ethanol, benzenemethanol, .alpha.-chloromethyl, benzenemethanol, a-chloromethyl, acmc-20apgz, benzyl alcohol, .alpha.-chloromethyl, acmc-20a5xf, 2-chloro-l-phenylethanol
PubChem CID 92898
IUPAC Name 2-chloro-1-phenylethanol
SMILES C1=CC=C(C=C1)C(CCl)O

Specifications

Density 1.193
Boiling Point 110°C to 112°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.55
Quantity 5 g
UN Number UN2810
Beilstein 907322
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 156.61
Percent Purity 97%
Chemical Name or Material (+/-)-2-Chloro-1-phenylethanol
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 216-816-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only