missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-methylimidazole, 97+%

Catalog No. AAH51867MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51867MD 250 mg
AAH5186703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51867MD Supplier Thermo Scientific Chemicals Supplier No. H51867MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 253453-91-7 | C4H5ClN2 | 116.55 g/mol

2-Chloro-1-methylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1-methylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 253453-91-7
Molecular Formula C4H5ClN2
Molecular Weight (g/mol) 116.55
MDL Number MFCD02179531
InChI Key VSCQFRRKWFQPOA-UHFFFAOYSA-N
PubChem CID 2773332
IUPAC Name 2-chloro-1-methylimidazole
SMILES CN1C=CN=C1Cl

Specifications

Melting Point -10°C
Quantity 250 mg
Synonym 2-chloro-1-methyl-1h-imidazole, 1h-imidazole, 2-chloro-1-methyl, pubchem7596, acmc-209gjb, 1-methyl-2-chloro-1h-imidazole, 2-chloro-n-methylimidazole, 1h-imidazole,2-chloro-1-methyl, 2-chloro-1n-methyl-imidazole, 2-chloro-1h-imidazole, n-me, 2-chloro-1h-imidazole, n-methyl
Formula Weight 116.55
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-1-methylimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only