missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,4-dimethoxybenzene, 99%

Catalog No. AAB2349718
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2349718 50 g
AAB2349730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2349718 Supplier Thermo Scientific Chemicals Supplier No. B2349718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2100-42-7 | C8H9ClO2 | 172.608 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2100-42-7
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.608
MDL Number MFCD00008356
InChI Key QMXZSRVFIWACJH-UHFFFAOYSA-N
Synonym 1-chloro-2,5-dimethoxybenzene, 2,5-dimethoxychlorobenzene, benzene, 2-chloro-1,4-dimethoxy, chlorohydroquinone dimethyl ether, chloro-1,4-dimethoxybenzene, 1,4-dimethoxy-2-chlorobenzene, o-chloro-p-dimethoxybenzene, monochloro-1,4-dimethoxybenzene, 6-chloro-1,4-dimethoxy benzene, acmc-209fgn
PubChem CID 246724
IUPAC Name 2-chloro-1,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)OC)Cl

Specifications

Density 1.21
Quantity 50 g
Refractive Index 1.547
Boiling Point 231°C to 234°C
Formula Weight 172.61
Percent Purity 99%
Chemical Name or Material 2-Chloro-1,4-dimethoxybenzene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-267-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only