missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-1,3-dimethylimidazolinium Chloride 98.0+%, TCI America™
Supplier: TCI America C14085G
Specifications
| 2-Chloro-1,3-dimethylimidazolinium Chloride | |
| White-Yellow | |
| MFCD09039290 | |
| 2-chloro-1,3-dimethylimidazolinium chloride, unii-56ka2a1u1d, 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium chloride, dmc mi, 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium chloride, pubchem16443, 2-chloro-1,3-dimethyl-4,5-dihydro-3h-imidazol-1-ium chloride, 1h-imidazolium, 2-chloro-4,5-dihydro-1,3-dimethyl-, chloride 1:1, 2-chloro-1,3-dimethylimidazolidin-2-ylium chloride | |
| [Cl-].CN1CC[N+](C)=C1Cl | |
| 169.05 | |
| 169.05 | |
| Crystalline Powder |
| 37091-73-9 | |
| C5H10Cl2N2 | |
| 5 g | |
| AEBBXVHGVADBHA-UHFFFAOYSA-M | |
| 2-chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium chloride | |
| 10176306 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 37091-73-9 | |
| 169.05 | |
| AEBBXVHGVADBHA-UHFFFAOYSA-M | |
| 10176306 | |
| [Cl-].CN1CC[N+](C)=C1Cl |
| C5H10Cl2N2 | |
| MFCD09039290 | |
| 2-chloro-1,3-dimethylimidazolinium chloride, unii-56ka2a1u1d, 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium chloride, dmc mi, 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium chloride, pubchem16443, 2-chloro-1,3-dimethyl-4,5-dihydro-3h-imidazol-1-ium chloride, 1h-imidazolium, 2-chloro-4,5-dihydro-1,3-dimethyl-, chloride 1:1, 2-chloro-1,3-dimethylimidazolidin-2-ylium chloride | |
| 2-chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium chloride |
Safety and Handling
TSCA : No