missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dimethylimidazolinium chloride, 90%

Catalog No. p-4229140
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439520050 5 g Glass bottle
AC439521000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439520050 Supplier Thermo Scientific Chemicals Supplier No. 439520050
Only null left

CAS: 37091-73-9 | C5H10Cl2N2 | 169.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 37091-73-9
Molecular Formula C5H10Cl2N2
Molecular Weight (g/mol) 169.05
MDL Number MFCD09039290
InChI Key AEBBXVHGVADBHA-UHFFFAOYSA-M
Synonym 2-chloro-1,3-dimethylimidazolinium chloride, unii-56ka2a1u1d, 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium chloride, dmc mi, 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium chloride, pubchem16443, 2-chloro-1,3-dimethyl-4,5-dihydro-3h-imidazol-1-ium chloride, 1h-imidazolium, 2-chloro-4,5-dihydro-1,3-dimethyl-, chloride 1:1, 2-chloro-1,3-dimethylimidazolidin-2-ylium chloride
PubChem CID 10176306
IUPAC Name 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium;chloride
SMILES [Cl-].CN1CC[N+](C)=C1Cl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 112°C to 121°C
Color White
Elemental Analysis 31.26 to 39.79% C, 14.58 to 18.56% N
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,3434
Formula Weight 169.05
Percent Purity 90%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-chloro-1, 3-dimethylimidazolinium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only