missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazolium Chloride 98.0+%, TCI America™

Catalog No. C34291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C34291G Supplier TCI America Supplier No. C34291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1228185-09-8
Molecular Formula C27H36Cl2N2
Molecular Weight (g/mol) 459.50
MDL Number MFCD23703069
InChI Key JDMACANGISWEGX-UHFFFAOYSA-M
PubChem CID 71463958
IUPAC Name 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2-chloro-3H-1λ⁵-imidazol-1-ylium chloride
SMILES [Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1Cl)C1=C(C=CC=C1C(C)C)C(C)C

Specifications

Melting Point 284°C
Color White-Yellow
Quantity 1 g
Formula Weight 459.50
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazolium Chloride