missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-1,10-phenanthroline 98.0+%, TCI America™
$318.75 - $1241.46
Chemical Identifiers
| CAS | 7089-68-1 |
|---|---|
| Molecular Formula | C12H7ClN2 |
| Molecular Weight (g/mol) | 214.65 |
| MDL Number | MFCD00185678 |
| InChI Key | JHRMQHFRVPVGHL-UHFFFAOYSA-N |
| PubChem CID | 355193 |
| IUPAC Name | 2-chloro-1,10-phenanthroline |
| SMILES | ClC1=CC=C2C=CC3=CC=CN=C3C2=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C1560100MG
|
TCI America
C1560100MG |
100 mg |
Each for $318.75
|
|
|||||
|
C15601G
|
TCI America
C15601G |
1 g |
Each for $1,241.46
|
|
|||||
Chemical Identifiers
| 7089-68-1 | |
| 214.65 | |
| JHRMQHFRVPVGHL-UHFFFAOYSA-N | |
| 2-chloro-1,10-phenanthroline |
| C12H7ClN2 | |
| MFCD00185678 | |
| 355193 | |
| ClC1=CC=C2C=CC3=CC=CN=C3C2=N1 |
Specifications
| 7089-68-1 | |
| White-Yellow | |
| MFCD00185678 | |
| JHRMQHFRVPVGHL-UHFFFAOYSA-N | |
| 2-chloro-1,10-phenanthroline | |
| 355193 | |
| ≥98.0% (HPLC,T) | |
| 2-Chloro-1,10-phenanthroline |
| 130°C | |
| C12H7ClN2 | |
| 100 mg | |
| ClC1=CC=C2C=CC3=CC=CN=C3C2=N1 | |
| 214.65 | |
| 214.65 | |
| Crystalline Powder |
Safety and Handling
TSCA : No