missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether, 97%

Catalog No. AAL1672506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1672506 5 g
AAL1672514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1672506 Supplier Thermo Scientific Chemicals Supplier No. L1672506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13838-16-9 | C3H2ClF5O | 184.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13838-16-9
Molecular Formula C3H2ClF5O
Molecular Weight (g/mol) 184.49
MDL Number MFCD00069095
InChI Key JPGQOUSTVILISH-UHFFFAOYNA-N
Synonym enflurane, methylflurether, efrane, ethrane, 2-chloro-1-difluoromethoxy-1,1,2-trifluoroethane, alyrane, anesthetic 347, 2-chloro-1,1,2-trifluoroethyl difluoromethyl ether, compound 347, anesthetic compound no. 347
PubChem CID 3226
ChEBI CHEBI:4792
IUPAC Name 2-chloro-1-(difluoromethoxy)-1,1,2-trifluoroethane
SMILES FC(F)OC(F)(F)C(F)Cl

Specifications

Density 1.517
Boiling Point 56°C to 57°C
Flash Point None
Refractive Index 1.303
Linear Formula CHF2OCF2CHClF
Quantity 5 g
Beilstein 1737129
Merck Index 14,3581
Formula Weight 184.49
Percent Purity 97%
Chemical Name or Material 2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-553-4
RTECSNumber KN6800000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only