missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxycinnamic acid, predominantly trans, 97%

Catalog No. AAL0146706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0146706 5 g
AAL0146714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0146706 Supplier Thermo Scientific Chemicals Supplier No. L0146706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 612-40-8 | C10H8O4 | 192.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 612-40-8
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00004380
InChI Key SCWPNMHQRGNQHH-AATRIKPKSA-N
Synonym 2-carboxycinnamic acid, 2-2-carboxyvinyl benzoic acid, 2-2-carboxyethenyl benzoic acid, e-2-2-carboxyvinyl benzoic acid, 2-e-3-hydroxy-3-oxoprop-1-enyl benzoic acid, 2-1e-2-carboxyvinyl benzoic acid, 2-e-2-carboxyethenyl benzoic acid, 2-1e-2-carboxyeth-1-en-1-yl benzoic acid, 2-e-2-carboxyvinyl benzoicacid, 2-carboxy cinnamic acid
PubChem CID 904938
IUPAC Name 2-[(E)-2-carboxyethenyl]benzoic acid
SMILES OC(=O)\C=C\C1=CC=CC=C1C(O)=O

Specifications

Melting Point 195°C to 205°C
Quantity 5 g
Beilstein 1870294
Formula Weight 192.17
Percent Purity 97%
Chemical Name or Material 2-Carboxycinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-308-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only