missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxybenzaldehyde, 98+%

Catalog No. AAA1120118
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1120118 50 g
AAA1120130 250 g
AAA112010B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1120118 Supplier Thermo Scientific Chemicals Supplier No. A1120118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 119-67-5 | C8H6O3 | 150.133 g/mol

2-Carboxybenzaldehyde is a carboxylated derivative of benzaldehyde which is readily converted to 2-hydroxymethyl benzoic acid by CBA dehydrogenase. 2-Carboxybenzaldehyde is a metabolite of ampicillin phthalidyl ester. It has been used to synthesize a series of N-substituted isoindolinones by reductive C-N coupling and intramolecular amidation with amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Carboxybenzaldehyde is a carboxylated derivative of benzaldehyde which is readily converted to 2-hydroxymethyl benzoic acid by CBA dehydrogenase. 2-Carboxybenzaldehyde is a metabolite of ampicillin phthalidyl ester. It has been used to synthesize a series of N-substituted isoindolinones by reductive C-N coupling and intramolecular amidation with amines.

Solubility
Soluble in water.

Notes
Air Sensitive. Store away from air and oxidizing agents. Incompatible with strong bases.

Chemical Identifiers

CAS 119-67-5
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00003336
InChI Key DYNFCHNNOHNJFG-UHFFFAOYSA-N
Synonym 2-carboxybenzaldehyde, phthalaldehydic acid, o-formylbenzoic acid, benzoic acid, 2-formyl, o-carboxybenzaldehyde, benzoic acid, formyl, benzaldehyde-2-carboxylic acid, formylbenzoic acid, carboxybenzaldehyde, pubchem7365
PubChem CID 8406
ChEBI CHEBI:17605
IUPAC Name 2-formylbenzoic acid
SMILES C1=CC=C(C(=C1)C=O)C(=O)O

Specifications

Melting Point 97°C to 101°C
Density 1.404
Flash Point 198°C (388°F)
Quantity 50 g
Beilstein 742381
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 150.13
Percent Purity ≥98%
Chemical Name or Material 2-Carboxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-342-3
RTECSNumber TH7015000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only