missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Carbobenzoxyamino)isobutyric Acid 98.0+%, TCI America™
$45.57 - $132.90
Chemical Identifiers
| CAS | 15030-72-5 |
|---|---|
| Molecular Formula | C12H15NO4 |
| Molecular Weight (g/mol) | 237.255 |
| MDL Number | MFCD00004190 |
| InChI Key | QKVCSJBBYNYZNM-UHFFFAOYSA-N |
| Synonym | z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl |
| PubChem CID | 294936 |
| IUPAC Name | 2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid |
| SMILES | CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 15030-72-5 | |
| 237.255 | |
| QKVCSJBBYNYZNM-UHFFFAOYSA-N | |
| 294936 | |
| CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
| C12H15NO4 | |
| MFCD00004190 | |
| z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl | |
| 2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid |
Specifications
| 79°C | |
| White | |
| MFCD00004190 | |
| QKVCSJBBYNYZNM-UHFFFAOYSA-N | |
| 2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid | |
| 237.255 | |
| 237.26 | |
| Crystalline Powder |
| 15030-72-5 | |
| C12H15NO4 | |
| z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl | |
| CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1 | |
| 5 g | |
| 294936 | |
| ≥98.0% (HPLC,T) | |
| 2-(Carbobenzoxyamino)isobutyric Acid |
Safety and Handling
TSCA : No