missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Carbobenzoxyamino)-2-hydroxyacetic Acid 98.0+%, TCI America™
$52.19 - $167.76
Chemical Identifiers
| CAS | 56538-57-9 |
|---|---|
| Molecular Formula | C10H11NO5 |
| Molecular Weight (g/mol) | 225.2 |
| MDL Number | MFCD02179279 |
| InChI Key | JQEZLSUFDXSIEK-UHFFFAOYSA-N |
| Synonym | 2-(Cbz-amino)-2-hydroxyacetic Acid, N-Carbobenzoxy-2-hydroxyglycine, Z-2-hydroxyglycine |
| PubChem CID | 3509506 |
| IUPAC Name | 2-hydroxy-2-(phenylmethoxycarbonylamino)acetic acid |
| SMILES | C1=CC=C(C=C1)COC(=O)NC(C(=O)O)O |
Chemical Identifiers
| 56538-57-9 | |
| 225.2 | |
| JQEZLSUFDXSIEK-UHFFFAOYSA-N | |
| 3509506 | |
| C1=CC=C(C=C1)COC(=O)NC(C(=O)O)O |
| C10H11NO5 | |
| MFCD02179279 | |
| 2-(Cbz-amino)-2-hydroxyacetic Acid, N-Carbobenzoxy-2-hydroxyglycine, Z-2-hydroxyglycine | |
| 2-hydroxy-2-(phenylmethoxycarbonylamino)acetic acid |
Specifications
| 189°C | |
| White | |
| MFCD02179279 | |
| JQEZLSUFDXSIEK-UHFFFAOYSA-N | |
| 2-hydroxy-2-(phenylmethoxycarbonylamino)acetic acid | |
| 225.2 | |
| 225.20 | |
| Crystalline Powder |
| 56538-57-9 | |
| C10H11NO5 | |
| 2-(Cbz-amino)-2-hydroxyacetic Acid, N-Carbobenzoxy-2-hydroxyglycine, Z-2-hydroxyglycine | |
| C1=CC=C(C=C1)COC(=O)NC(C(=O)O)O | |
| 1 g | |
| 3509506 | |
| ≥98.0% (HPLC,T) | |
| 2-(Carbobenzoxyamino)-2-hydroxyacetic Acid |
Safety and Handling
TSCA : No