missing translation for 'onlineSavingsMsg'
Learn More

2-Butyne 97.0+%, TCI America™

Catalog No. B09515ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09515ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B09515ML Supplier TCI America Supplier No. B09515ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 503-17-3
Molecular Formula C4H6
Molecular Weight (g/mol) 54.092
MDL Number MFCD00009275
InChI Key XNMQEEKYCVKGBD-UHFFFAOYSA-N
Synonym 2-butyne, dimethylacetylene, crotonylene, unii-lke6d3018e, ch3cequivcch3, 1,2-dimethylvinylene, 1-methyl-1-propynyl, 2-butyn, acmc-209kl9
PubChem CID 10419
IUPAC Name but-2-yne
SMILES CC#CC

Specifications

Color Colorless
Boiling Point 27°C
Quantity 5 mL
UN Number 1144
Formula Weight 54.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Butyne
EINECSNumber (2)-3161
RTECSNumber GQ7210000
TSCA Yes
Recommended Storage Refrigerator