missing translation for 'onlineSavingsMsg'
Learn More

2-Butyne, 98%, Thermo Scientific Chemicals

Catalog No. p-3211627
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC135140050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC135140050 Supplier Thermo Scientific Chemicals Supplier No. 135140050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 503-17-3
Molecular Formula C4H6
Molecular Weight (g/mol) 54.09
MDL Number MFCD00009275
InChI Key XNMQEEKYCVKGBD-UHFFFAOYSA-N
Synonym 2-butyne, dimethylacetylene, crotonylene, unii-lke6d3018e, ch3cequivcch3, 1,2-dimethylvinylene, 1-methyl-1-propynyl, 2-butyn, acmc-209kl9
PubChem CID 10419
IUPAC Name but-2-yne
SMILES CC#CC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -32°C
Color Colorless to Yellow
Density 0.6900g/mL
Boiling Point 24°C to 27°C
Flash Point -25°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3920 to 1.394
Linear Formula CH3CCCH3
Quantity 5 g
Beilstein 01, 249
Specific Gravity 0.69
Formula Weight 54.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Butyne
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Extremely flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-962-2

RUO – Research Use Only