missing translation for 'onlineSavingsMsg'
Learn More

2-Butyne-1,4-diol 99.0+%, TCI America™

Catalog No. p-7118656
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
B074925G 25 g
B0749500G 500 g
2 options

Catalog No. B074925G

Supplier: TCI America B074925G

Only null left

Chemical Identifiers

CAS 110-65-6
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002915
InChI Key DLDJFQGPPSQZKI-UHFFFAOYSA-N
Synonym 2-butyne-1,4-diol, butynediol, 1,4-dihydroxy-2-butyne, 1,4-butynediol, bis hydroxymethyl acetylene, 2-butynediol, 2-butin-1,4-diol, agrisynth b3d, unii-axh202fpqm, 1,4-butynediol van
PubChem CID 8066
ChEBI CHEBI:16413
IUPAC Name but-2-yne-1,4-diol
SMILES C(C#CCO)O

Specifications

Melting Point 58°C
Boiling Point 238°C
Quantity 25 g
UN Number 2716
Formula Weight 86.09
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butyne-1,4-diol
EINECSNumber (2)-0268
RTECSNumber ES0525000
TSCA Yes