missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 2-Butyn-1-ol | |
| Yellow | |
| C4H6O | |
| 5 mL | |
| 2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol | |
| CC#CCO | |
| 70.091 | |
| 70.09 | |
| Liquid |
| 764-01-2 | |
| 145°C | |
| MFCD00002914 | |
| 1987 | |
| NEEDEQSZOUAJMU-UHFFFAOYSA-N | |
| but-2-yn-1-ol | |
| 12991 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 764-01-2 | |
| 70.091 | |
| NEEDEQSZOUAJMU-UHFFFAOYSA-N | |
| 12991 | |
| CC#CCO |
| C4H6O | |
| MFCD00002914 | |
| 2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol | |
| but-2-yn-1-ol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator